Learn More
Thermo Scientific™ EZ-Link™ Maleimide-PEG11-Biotin
Description
Thermo Scientific™ EZ-Link Maleimide-PEG11-Biotin is a long, maleimide-activated, sulfhydryl-reactive biotinylation reagent that includes an 11-unit polyethylene glycol spacer arm for increased water-solubility and reach.
Highlights:
- Polyethylene glycol (PEG) spacer yields biotin-labeled proteins with greater solubility and less aggregation
- Forms stable thioether bond at pH 6.5-7.5
- PEG spacer improves aqueous solubility, allowing labeling to be performed under non-denaturing, physiological conditions
- Extended spacer arm reduces steric hindrance in binding to avidin, streptavidin and Thermo Scientific NeutrAvidin Proteins
Specifications
Specifications
| Product Type | Maleimide-PEG11-Biotin |
| Quantity | 25 mg |
| Content And Storage | Store desiccated at 4°C. Shipped at ambient temperature. |
| Product Line | EZ-Link |
| Cell Permeability | Cell-Impermeant |
| Chemical Reactivity | Thiol |
| Reactive Moiety | Maleimide |
| Solubility | DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water |
| Label or Dye | Biotin |
| Spacer | Extra-long, Pegylated |
| Show More |
Frequently Asked Questions (FAQs)
Both maleimide- and iodoacetyl-containing biotinylation reagents react with sulfhydryl (-SH) groups to form stable thioether bonds that are not cleavable. EZ-Link HPDP-Biotin on the other hand is a pyridyldithiol-biotin compound that reacts with -SH groups in near-neutral buffers to form reversible disulfide bonds. Because the disulfide group can be cleaved using DTT or other reducing agents, HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule.
For Research Use Only. Not for use in diagnostic procedures.