missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Iodoacetyl-PEG2-Biotin

Catalog No. PI21334
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21334 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21334 Supplier Thermo Scientific™ Supplier No. 21334

Biotinylate reduced cysteines (-SH groups) at high pH with this haloacetyl-activated, sulfhydryl-reactive biotinylation reagent containing polyethylene glycol for solubility.

Thermo Scientific™ EZ-Link Iodoacetyl-PEG2-Biotin is a mid-length, haloacetyl-activated, sulfhydryl-reactive biotinylation reagent that contains a 2-unit ethylene glycol in its spacer arm for increased water-solubility characteristics.

Highlights:

  • Water-soluble, polyethylene glycol (PEG) spacer arm
  • Reacts with sulfhydryl (-SH) groups at basic to neutral pH
  • Coupling reactions occur in the dark
  • PEG spacer arm transfers hydrophilicity to final conjugate
  • Forms a stable thioether bond

Specifications

Product Type Iodoacetyl-PEG2-Biotin
Quantity 50 mg
Content And Storage Store desiccated at 4°C. Shipped at ambient temperature.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Thiol
Reactive Moiety Haloacetyl
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide), Water
Label or Dye Biotin
Spacer Mid-length, Pegylated
Label Type Biotin & Analogs
Show More Show Less
What is the advantage of using EZ-Link HPDP-Biotin over maleimide- or iodoacetyl-containing biotinylation reagents?

Both maleimide- and iodoacetyl-containing biotinylation reagents react with sulfhydryl (-SH) groups to form stable thioether bonds that are not cleavable. EZ-Link HPDP-Biotin on the other hand is a pyridyldithiol-biotin compound that reacts with -SH groups in near-neutral buffers to form reversible disulfide bonds. Because the disulfide group can be cleaved using DTT or other reducing agents, HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule.

For Research Use Only. Not for use in diagnostic procedures.