missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ HPDP-Biotin, No-Weigh™ Format

Catalog No. p-7180329
Change view
Click to view available options
Quantity:
50 mg
10 x 1 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21341 50 mg
PIA35390 10 x 1 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. PI21341 Supplier Thermo Scientific™ Supplier No. 21341
Only null left

Thermo Scientific™ EZ-Link HPDP-Biotin is pyridyldithiol-activated, sulfhydryl-reactive biotinylation reagent that conjugates via a cleavable (reversible) disulfide bond to enable use in a variety of purification methods.

Highlights:

  • Pyridyldithiol reacts with -SH groups to form a stable disulfide bond
  • Absorbance of pyridine-2-thione leaving group at 343nm can be used to determine reaction progress
  • Disulfide bond is cleavable using 50mM DTT (Part No. 20291) or 100mM β-Mercaptoethanol (Part No. 35600)
  • Extended spacer arm prevents steric hindrance
  • Must be dissolved in DMSO or DMF before use
  • Biotinylation occurs between pH 6-9

Specifications

Product Type HPDP-Biotin
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Thiol
Reactive Moiety Pyridyldisulfide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Long, Cleavable
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.