missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ BMCC-Biotin

Catalog No. pi21900
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21900 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21900 Supplier Thermo Scientific™ Supplier No. LSG21900

Biotinylate reduced cysteines (-SH groups) with a maleimide-activated, sulfhydryl-reactive biotinylation reagent containing a stabilizing cyclohexane group in the spacer arm.

Thermo Scientific™ EZ-Link BMCC-Biotin is a maleimide-activated, sulfhydryl-reactive biotinylation reagent with an extended spacer arm that contains a stabilizing cyclohexane group.

Highlights:

  • Biotinylates at pH 6.5-7.5
  • Reaction avoids potential modification of tyrosine groups that can inactivate biological activity
  • Longer spacer arm than iodoacetyl biotin
  • Must be dissolved in DMSO or DMF before use

Specifications

Product Type BMCC-Biotin
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Permeant
Chemical Reactivity Thiol
Reactive Moiety Maleimide
Solubility DMF (Dimethylformamide), DMSO (Dimethylsulfoxide)
Label or Dye Biotin
Spacer Extra-long
Label Type Biotin & Analogs
Show More Show Less
What is the advantage of using EZ-Link HPDP-Biotin over maleimide- or iodoacetyl-containing biotinylation reagents?

Both maleimide- and iodoacetyl-containing biotinylation reagents react with sulfhydryl (-SH) groups to form stable thioether bonds that are not cleavable. EZ-Link HPDP-Biotin on the other hand is a pyridyldithiol-biotin compound that reacts with -SH groups in near-neutral buffers to form reversible disulfide bonds. Because the disulfide group can be cleaved using DTT or other reducing agents, HPDP-Biotin is useful for labeling and affinity-purification applications that require recovery of the original, unmodified molecule.

For Research Use Only. Not for use in diagnostic procedures.