missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ EZ-Link™ Amine-PEG2-Biotin

Catalog No. pi21346
Change view
Click to view available options
Quantity:
50 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PI21346 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PI21346 Supplier Thermo Scientific™ Supplier No. LSG21346

Biotinylate proteins or surfaces using EDC or other amine-reactive crosslinkers with this short (20.4A) biotinylation reagent containing a 2-unit polyethylene glycol spacer.

Thermo Scientific™ EZ-Link Amine-PEG2-Biotin is a biotinylation reagent that contains a 2-unit polyethylene glycol (PEG) spacer arm and a terminal primary amine for conjugation and protein labeling with EDC or other crosslinkers.

Amine-PEG2-Biotin and is a water-soluble biotin compound containing a polyethylene glycol (PEG) spacer arm and a terminal primary amine. The hydrophilic PEG spacer arm imparts water solubility that is transferred to molecules that are conjugated to the biotin compound. The PEG spacer arm also gives this reagent a long and flexible connection to minimize steric hindrance involved with binding to avidin molecules. The amine group can be reacted with the crosslinker EDC to form conjugates with carboxylates and 5' phosphates.

Highlights:

  • Use the EDC crosslinker to couple this analog to carboxyl (-COOH) groups or 5' phosphate groups
  • Water-soluble, polyethylene glycol (PEG) spacer arm
  • PEG spacer arm transfers hydrophilicity to final conjugate

Specifications

Product Type Amine-PEG2-Biotin
Quantity 50 mg
Content And Storage Store at 4°C.
Product Line EZ-Link
Cell Permeability Cell-Impermeant
Chemical Reactivity Carboxylic Acid, Ketone, Aldehyde
Reactive Moiety Amine
Solubility Water
Label or Dye Biotin
Spacer Mid-length, Pegylated
Label Type Biotin & Analogs
Show More Show Less

For Research Use Only. Not for use in diagnostic procedures.