Learn More
MilliporeSigma EZ-Fit™ Manifold
3-place manifold For EZ-Fit™ filtration units, made out of stainless steel
Supplier: MilliporeSigma EZFITSAM3
Description
EZ-Fit™ Filtration Manifold Base with 3 places is uniquely designed to carry out laboratory filtration of samples. The filtration manifold construction eliminates all sources of contamination over repeated use as each part of the manifold base is cleanable and autoclavable.
The EZ-Fit™ Filtration Manifold Base can accommodate three filtration heads at a time, which would mean three samples can be tested at one go, and the ergonomic design enables operation under laminar flow hood and save laboratory bench space.
The 2-way valve system of the filtration manifold prevents the user to accidentally touch the filtration head while opening or closing the valve.
Specifications
| Reusable | |
| Stainless Steel | |
| Manifold |
| EZ-Fit Filtration Manifold | |
| Non-sterile |