Learn More
MilliporeSigma™ Extran™ MA 02
Use for manual cleaning. MilliporeSigma Extran™ MA 02 is a pH-neutral concentrated liquid that contains phosphates. Extran cleaning agents have a consistent composition and are biodegradable, with no residues, impurities, harsh chemicals or scents. (
Supplier:  MilliporeSigma™ 1.07553.5000
 View more versions of this product
Catalog No. M1075535000
Only null left
Description
Description
This product is identical to EX0992 with the same quality and specification. Some pack sizes have changed to provide a globally harmonized offering.
Specifications
Chemical Identifiers
Concentrated Liquid
Colorless
Plastic
Manual cleaning
6 to 8 (5%, aq. soln.)
100°C
1.07 g/cm3 (20°C)
Specifications
Concentrated Liquid
Colorless
Plastic
Manual cleaning
6 to 8 (5%, aq. soln.)
100°C
1.07 g/cm3 (20°C)
5 L
Product Suggestions
Product Suggestions