missing translation for 'onlineSavingsMsg'
Learn More

Extraction Medium Stock Solution, MilliporeSigma™ Supelco™

Catalog No. 111016744
Change view
Click to view available options
Quantity:
100 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111016744 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111016744 Supplier Merck Emd Millipore Supplier No. 49574100MLF

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Featured Industries: Environmental

Specifications

CAS 142-82-5
Flash Point -4°C - closed cup
Type Standard
Quantity 100 mL