missing translation for 'onlineSavingsMsg'
Learn More

Velp™ Extraction Cup for SER158/3 Solvent Extractor

Catalog No. 11392077
Change view
Click to view available options
Quantity:
3 Cups
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
11392077 3 Cups
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 11392077 Supplier Velp™ Supplier No. A00000290

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Extraction cup for use with the SER158/3 Solvent Extractor.

  • Standard extraction cup for solvent extractor

Specifications

For Use With (Equipment) SER 158/3 Solvent Extractor
Product Type Extraction Cup
Sterility Non-sterile
Color Clear
Quantity 3 Cups
Diameter (Metric) 56 mm
Height (Metric) 120 mm
Voltage 115/230 VAC