Learn More
Apex Aseptic Extra-Safe™ Lab Jackets, White
Static-free unisex lab coat is made with a 3-layer SMS material to provide protection and comfort in a variety of environments.
$59.09 - $74.85
Specifications
| Product Type | General Purpose Lab Coat |
|---|---|
| Gender | Unisex |
| Material | SMS |
| Color | White |
| Closure Type | Snaps |
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Size | Price | Quantity | |||||
|
22770000
|
Apex Aseptic
3630WHXS |
X-Small |
Pack of 10 Each for $59.09
Case of 10 Pack for $560.40
|
|
|||||
|
22770001
|
Apex Aseptic
3630WHS |
Small |
Pack of 10 Each for $59.09
Case of 10 Pack for $560.40
|
|
|||||
|
22770002
|
Apex Aseptic
3630WHM |
Medium |
Pack of 10 Each for $59.09
Case of 10 Pack for $560.40
|
|
|||||
|
22770003
|
Apex Aseptic
3630WHL |
Large |
Pack of 10 Each for $59.09
Case of 10 Pack for $560.40
|
|
|||||
|
22770004
|
Apex Aseptic
3630WHXL |
X-Large |
Pack of 10 Each for $59.09
Case of 10 Pack for $560.40
|
|
|||||
|
22770005
|
Apex Aseptic
3630WH2XL |
2X-Large |
Pack of 10 Each for $63.04
Case of 10 Pack for $597.80
|
|
|||||
|
22770006
|
Apex Aseptic
3630WH3XL |
3X-Large |
Pack of 10 Each for $66.98
Case of 10 Pack for $635.20
|
|
|||||
|
22770007
|
Apex Aseptic
3630WH4XL |
4X-Large |
Pack of 10 Each for $70.93
Case of 10 Pack for $672.60
|
|
|||||
|
22770008
|
Apex Aseptic
3630WH5XL |
5X-Large |
Pack of 10 Each for $74.85
Case of 10 Pack for $709.80
|
|
|||||
Description
- Made of soft three-layer SMS (Spunbond/Meltblown/Spunbond) fabric
- Fluid-resistant, breathable
- Snap front, three pockets, knit cuffs and collar
- Hip length
- Autoclavable and washable up to five times
- Latex-free
Specifications
| General Purpose Lab Coat | |
| SMS | |
| Snaps | |
| Limited-use | |
| Fluids | |
| 3 | |
| Yes | |
| 3-ply |
| Unisex | |
| White | |
| Knit | |
| Non-sterile | |
| Hip-length | |
| Knit | |
| 3630WH | |
| Not Autoclavable |