Learn More
Extension Clamps
Allow for variable distance between the object and support rod.
Supplier:  Customer Channels Group (Ccg) HUMH8572
 View more versions of this product
Catalog No. S49099
Only null left
Description
Description
  • Plastic-covered jaws hold beakers and flasks firmly at any angle at a variable distance from their support
  • One jaw is fixed; the other opens against a stainless-steel compression spring
  • Open gradually to maximum size without binding
  • Clamps will not rust
Specifications
Specifications
Extension Clamp
9.05 in.
9 in. Extension Clamp 2 Prong
Steel, Plastisol
22.98 cm
Product Suggestions
Product Suggestions