missing translation for 'onlineSavingsMsg'
Learn More

OHAUS EXPLORER PLUS™ Balance

Catalog No. 01185032
Change view
Click to view available options
Capacity:
120 g
120.0 g
220 g
220.0 g
320.0 g
420.0 g
620.0 g
1200.0 g
2200.0 g
4200.0 g
6200.0 g
8200.0 g
10200.0 g
12200.0 g
24000 g
35000 g
65000 g
Readability:
0.01 mg
0.1 mg
1 mg
0.01 g
0.01g
0.1 g
22 product options available for selection
Product selection table with 22 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity Readability
01185032 120.0 g 0.1 mg
01185053 10200.0 g 0.1 g
01185034 1200.0 g 1 mg
01185063 12200.0 g 0.01 g
01185060 120.0 g 0.01 mg
01185071 120 g 0.01 mg
01185051 2200.0 g 0.01 g
01185049 220.0 g 1 mg
01185048 220.0 g 0.1 mg
01185062 220.0 g 0.01 mg
01185061 220 g 0.01 mg
01185037 24000 g 0.1 g
01185059 320.0 g 0.1 mg
01185065 35000 g 0.1 mg
01185035 4200.0 g 0.01g
01185050 420.0 g 1 mg
01185036 6200.0 g 0.1 g
01185052 6200.0 g 0.01 g
01185033 620.0 g 1 mg
01185038 65000 g 0.1 mg
01185064 8200.0 g 0.1 g
01185072 8200.0 g 0.01g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 22 options available.
22 options
Catalog No. 01185032 Supplier OHAUS Supplier No. 31059173

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

The new Explorer EXP series offers a premium weighing performance, compliance support and data traceability

  • Seamless setup with motorized leveling, anti-pintch auto doors, touchless sensors, ionizers and draft shield lights. Has a 7 in. glass touchscreen, chemical protection and 2 modifiable status lights
  • Ensure compliance with 4 level user system, password protection, and 100k internal system log
  • The time synchronization NTP function confirms data traceability to meet the Pharmaceutical regulations
  • Advanced communication capabilities with straightforward integration to Laboratory
  • Information Management Systems (LIMS), and hassle-free data transfer with driverless HID for easy data input

Specifications

Capacity 120.0 g
Display Type Touchscreen
Readability 0.1 mg
Type Analytical Balances