missing translation for 'onlineSavingsMsg'
Learn More

Gibco™ ExpiCHO™ Stable Production Medium

Catalog No. A3711001
Change view
Click to view available options
Quantity:
1000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A3711001 1000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. A3711001 Supplier Gibco™ Supplier No. A3711001

missing translation for 'validMainframeMsg'

Gibco ExpiCHO Stable Production Medium (SPM) is specifically designed to simplify your cell line development process into production with minimal effort and a high degree of confidence.

This chemically defined, protein-free, animal origin component-free formulation is fully optimized for large-scale fed-batch culture with ExpiCHO-S cells. ExpiCHO SPM supports an easy transition from transient to stable production, provides high titers without the need for additional medium optimization (see figures below), and works well with existing EfficientFeed C+ supplement.

ExpiCHO SPM is part of the ExpiCHO family, an end-to-end solution that empowers you to carry out your entire workflow from transient to stable expression without having to screen or optimize media and feeds.

Hassle-free scale-up
Simplify your process as you transition into stable clones with ExpiCHO SPM—no cell adaptation, no media optimization, no changes in cell doubling time and peak density. ExpiCHO SPM eases the transition using ExpiCHO-S cells and provides optimal performance with less need for fine-tuning from development scientists to get you to the next step faster. This medium supports high titers under standard fed-batch process conditions in conjunction with EfficientFeed C+ supplement. It is available in liquid format (this page) or in our AGT dry format from 10 L through 450 L equivalents with larger sizes available upon request.

ONE Solution
The ExpiCHO family includes products that are engineered to work seamlessly with each other. ExpiCHO SPM is a member of this family and allows expanded support of bioproduction from transient to stable protein expression. Look for ExpiCHO-branded protocols and methods to get optimal performance in the development process.

Members of the ExpiCHO family include:
• ExpiCHO Stable Production Medium (this page)
• ExpiCHO Expression Medium
• ExpiCHO-S cells
• Expifectamine CHO Transfection Kit

The power to streamline transfer to manufacturing
ExpiCHO SPM liquid format and AGT dry format demonstrate comparable cell growth and titer performance (see figure below). Therefore, scale-up and manufacturing using AGT dry format is simplified by knowing that the reconstituted AGT dry format will behave similarly to the liquid format used prior to scale-up. The AGT dry format is easy to reconstitute; just add water and it will automatically adjust pH and osmolality. This can help provide efficiencies in supply chain complexity, reconstitution complexity, and, therefore, overall expense.

Specifications

Cell Line ExpiCHO-S
Classification Animal Origin-free, Chemically-defined, Protein-free
Form Liquid
Product Type Stable Production Medium (SPM)
Product Line ExpiCHO
Quantity 1000 mL
Shipping Condition Room Temperature
Format Bottle
Content And Storage Store in refrigerator (2°C to 8°C).

For Research Use Only. Not for use in diagnostic procedures.