missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ EXOC3 Recombinant Rabbit Monoclonal Antibody (23GB5345)

Catalog No. MA552841
Change view
Click to view available options
Quantity:
20 μL
100 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA552841 20 μL
MA552840 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MA552841 Supplier Invitrogen™ Supplier No. MA552841
Only null left

Rabbit Recombinant Monoclonal Antibody

EXOC3 Recombinant Monoclonal Antibody for Flow, ICC/IF, Western Blot

The protein encoded by this gene is a component of the exocyst complex, a multiple protein complex essential for targeting exocytic vesicles to specific docking sites on the plasma membrane. Though best characterized in yeast, the component proteins and functions of exocyst complex have been demonstrated to be highly conserved in higher eukaryotes. At least eight components of the exocyst complex, including this protein, are found to interact with the actin cytoskeletal remodeling and vesicle transport machinery. The complex is also essential for the biogenesis of epithelial cell surface polarity.

Specifications

Antigen EXOC3
Applications Flow Cytometry, Western Blot, Immunocytochemistry
Classification Recombinant Monoclonal
Clone 23GB5345
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene EXOC3
Gene Accession No. O60645, Q62825, Q6KAR6
Gene Alias 2810050O03Rik; E430013E20Rik; EXOC3; EXOC3L; EXOC3L1; Exocyst complex component 3; exocyst complex component 3 like 1; exocyst complex component 3-like 1; exocyst complex component 3-like protein; Exocyst complex component Sec6; I79_024581; KIAA0895L; KIAA0895-like ortholog; LIM/homeobox protein Lhx2 pseudogene; protein Jiangli; RGD1311132; rSec6; Sec 6 homolog; Sec6; Sec6 protein; SEC6L1; SEC6-like 1; Sec6p
Gene Symbols EXOC3
Host Species Rabbit
Immunogen A synthesized peptide derived from human EXOC3 (680-700AA).
Purification Method Affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 11336, 211446, 252881
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less