missing translation for 'onlineSavingsMsg'
Learn More

exo-Norborneol 98.0+%, TCI America™

Catalog No. p-7133968
Change view
Click to view available options
Quantity:
5 g
25 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
N07755G 5 g
N077525G 25 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. N07755G Supplier TCI America Supplier No. N07755G
Only null left

Chemical Identifiers

CAS 497-37-0
Molecular Formula C7H12O
Molecular Weight (g/mol) 112.17
MDL Number MFCD00167747,MFCD00136051
InChI Key ZQTYQMYDIHMKQB-UHFFFAOYNA-N
PubChem CID 12313473
IUPAC Name bicyclo[2.2.1]heptan-2-ol
SMILES OC1CC2CCC1C2

Specifications

Melting Point 128°C
Color White
Boiling Point 177°C
Quantity 5 g
Formula Weight 112.17
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material exo-Norborneol