missing translation for 'onlineSavingsMsg'
Learn More
Learn More
exo-Norborneol 98.0+%, TCI America™
Supplier: TCI America N077525G
Specifications
| exo-Norborneol | |
| 128°C | |
| 177°C | |
| MFCD00167747,MFCD00136051 | |
| ZQTYQMYDIHMKQB-UHFFFAOYNA-N | |
| bicyclo[2.2.1]heptan-2-ol | |
| 12313473 | |
| ≥98.0% (GC) |
| 497-37-0 | |
| White | |
| C7H12O | |
| 25 g | |
| OC1CC2CCC1C2 | |
| 112.17 | |
| 112.17 | |
| Crystalline Powder |
Chemical Identifiers
| 497-37-0 | |
| 112.17 | |
| ZQTYQMYDIHMKQB-UHFFFAOYNA-N | |
| bicyclo[2.2.1]heptan-2-ol |
| C7H12O | |
| MFCD00167747,MFCD00136051 | |
| 12313473 | |
| OC1CC2CCC1C2 |
Safety and Handling
TSCA : Yes