Learn More
Exemestane Related Compound D Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™
Description
Exemestane Related Compound D is an impurity of the steroidal anticancer drug exemestane. Exemestane belongs to the class of antiestrogens known as aromatase inhibitors and it is commonly used for the treatment of breast cancer.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.
Specifications
Specifications
| Purity Grade Notes | These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available. |
| Type | Certified Reference Material |
| Quantity | 30 mg |
| Grade | Certified Reference Material |