missing translation for 'onlineSavingsMsg'
Learn More

Exemestane Related Compound A Pharmaceutical Secondary Standard, MilliporeSigma™ Supelco™

Catalog No. 111013433
Change view
Click to view available options
Quantity:
30 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
111013433 30 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 111013433 Supplier Merck Emd Millipore Supplier No. PHR189430MG
Exemestane Related Compound A is an impurity of the steroidal anticancer drug, exemestane. Exemestane belongs to the class of antiestrogens known as aromatase inhibitors. It is commonly used for the treatment of breast cancer.Pharmaceutical secondary standards for application in quality control, provide pharma laboratories and manufacturers with a convenient and cost-effective alternative to the preparation of in-house working standards.

Specifications

CAS 19457-55-7
Purity Grade Notes These secondary standards offer multi-traceability to the USP, EP and BP primary standards, where they are available.
Linear Formula C20H26O2
Synonym 6-Methyleneandrost-4-ene-3,17-dione
Type Certified Reference Material
Quantity 30 mg
Formula Weight 298.42
Grade Certified Reference Material