Learn More
Air-Tite Exel Syringe
These syringes are latex free, sterile, non-pyrogenic and non-toxic.
Supplier: Air-Tite EL20
Description
The design includes:
- Bold printing
- Clear graduations
- Siliconized gaskets for smooth, consistent plunger motion
- A positive plunger stop
- FDA registered
- CE and ISO 9001 certified
Specifications
| Not Autoclavable | |
| No Needle Included | |
| Disposable Syringe | |
| 20 to 24 mL | |
| Centric Luer Lock | |
| Sterile |
| Polypropylene, Silicone Oil (Lubricant) | |
| Yes | |
| Single-use | |
| Hypodermic Needle, Safety Needle, Blunt Fill Needle | |
| Yes |