Learn More
Excelta™ Small Parts Handling Tweezers
Choice of small, round, broad angle, broad and curved flat tips
Supplier:  Excelta™ 36ASA
 View more versions of this product
Catalog No. 17467253
Only null left
Description
Description
  • Available in high and general precision
  • Made of carbon tool steel, 300 and 400 series stainless-steel
  • Choice of small, round, broad angle, broad and curved flat tips
Specifications
Chemical Identifiers
Stainless Steel
Each
0.038 cm
0.81 cm
Tweezers
0.015 in.
0.32 in.
Specifications
Offset Broad Flat Tip
Stainless Steel
Tweezers
Ideal for picking flat objects off a flat surface, serrated handles for better control
0.038 cm
0.32 in.
4.75 in.
120 mm
Each
0.015 in.
Three Star High
0.81 cm
Product Suggestions
Product Suggestions