Learn More
Excelta™ Roto-Pic Vacuum Systems
For vacuum pick-up of chips, wafers
$3,213.44 - $8,310.28
Specifications
For Use With (Application) For vacuum pickup of chips and wafers
View More Specs
Products 2
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
Catalog Number Mfr. No. Product Type Price Quantity & Availability  
17468535
View Documents Promotion Details
Excelta™
2000R
Vacuum System
Each for $3,213.44
Only null left
17468536
View Documents Promotion Details
Excelta™
2000RK
Vacuum Pickup Kit
Each for $8,310.28
Only null left
Description
Description
  • Includes handpiece, plastic hose, vacuum pump
  • Positive hold-and-release slide bar, 360° rotatable nose wheel
  • Capacity: 6 in. (15.2mm) of Mercury
  • Complete Vacuum Kit (Mfr. No. 2000RK) includes Probe Kit (Mfr. No. 2000P), Vacuum Cup Kit (Mfr. No. 2000WP), Vacuum Tip Kit (Mfr. No. 2000DA)
Specifications
Specifications
For vacuum pickup of chips and wafers