Learn More
Excelta™ Roto-Pic Vacuum Systems
Catalog No. 17468535
Change view
Click to view available options
Product Type:
Vacuum Pickup Kit
Vacuum System
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Product Type
17468535 Vacuum System
17468536 Vacuum Pickup Kit
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 17468535 Supplier Excelta™ Supplier No. 2000R
For vacuum pick-up of chips, wafers
  • Includes handpiece, plastic hose, vacuum pump
  • Positive hold-and-release slide bar, 360° rotatable nose wheel
  • Capacity: 6 in. (15.2mm) of Mercury
  • Complete Vacuum Kit (Mfr. No. 2000RK) includes Probe Kit (Mfr. No. 2000P), Vacuum Cup Kit (Mfr. No. 2000WP), Vacuum Tip Kit (Mfr. No. 2000DA)
Specifications
Product Type Vacuum System
For Use With (Application) For vacuum pickup of chips and wafers
Includes Hand piece, Plastic hose, vacuum pump