missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ EVPL Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5150267
Description
EVPL Polyclonal Antibody for Western Blot, ICC/IF, IHC
This gene encodes a member of the DEAH-containing family of RNA helicases. The encoded protein is an enzyme that catalyzes the ATP-dependent unwinding of double-stranded RNA and DNA-RNA complexes. This protein localizes to both the nucleus and the cytoplasm and functions as a transcriptional regulator. This protein may also be involved in the expression and nuclear export of retroviral RNAs. Alternate splicing results in multiple transcript variants. Pseudogenes of this gene are found on chromosomes 11 and 13.Chemical Identifiers
| EVPL | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92817 | |
| 2125 | |
| Recombinant Envoplakin (EVPL) (Lys3-Met258). | |
| Antibody |
| 0.5 mg/mL | |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| Unconjugated | |
| Rabbit | |
| Human | |
| EVPL | |
| 210 kDa cornified envelope precursor protein; 210 kDa paraneoplastic pemphigus antigen; 210-kD protein; 210kDa protein; envoplakin; EVPK; Evpl; p210 | |
| EVPL | |
| Primary | |
| Protein A |
Specifications
| EVPL | |
| Polyclonal | |
| Unconjugated | |
| EVPL | |
| 210 kDa cornified envelope precursor protein; 210 kDa paraneoplastic pemphigus antigen; 210-kD protein; 210kDa protein; envoplakin; EVPK; Evpl; p210 | |
| Rabbit | |
| Protein A | |
| RUO | |
| 2125 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry, Western Blot, Immunocytochemistry | |
| 0.5 mg/mL | |
| PBS with 50% glycerol and 0.05% ProClin 300; pH 7.4 | |
| Q92817 | |
| EVPL | |
| Recombinant Envoplakin (EVPL) (Lys3-Met258). | |
| 200 μL | |
| Primary | |
| Human | |
| Antibody |