missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Budigalimab (PDCD1/PD1/CD279) - Research Grade Biosimilar, ProSci
Description
Budigalimab (PDCD1/PD1/CD279) Recombinant Monoclonal Antibody specifically detects Budigalimab (PDCD1/PD1/CD279).
Specifications
Specifications
| Antigen | Budigalimab (PDCD1/PD1/CD279) |
| Applications | Not Available |
| Classification | Recombinant Monoclonal |
| Conjugate | Unconjugated |
| Formulation | PBS buffer pH7.5 |
| Gene Alias | ABBV-181,PR-1648817 |
| Host Species | Chinese Hamster (CHO) |
| Purification Method | >95% |
| Quantity | 0.1 mg |
| Regulatory Status | RUO |
| Show More |