Learn More
Everolimus
A rapamycin analog | CAS: 159351-69-6 | C53H83NO14 | 958.24 g/mol
Supplier: Thermo Scientific Chemicals J60139MA
Description
Everolimus is a rapamycin analog. It is used to treat certain types of kidney cancer, breast cancer and brain tumor. It prevents transplant rejection by decreasing the activity of the immune system. It also act as a kinase inhibitor. It is used in the treatment of subependymal giant cell astrocytoma (SEGA; a type of brain tumor) in adults and children 1 year of age and older who have TSC.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
SolubilitySoluble in dimethysulfoxide,ethanol and chloroform. Slightly soluble in water.
Notes
Incompatible with strong oxidizing agents.
Specifications
| Everolimus | |
| White | |
| MFCD07785165 | |
| 14,3907 | |
| HKVAMNSJSFKALM-PWMUVMERSA-N | |
| 958.24 | |
| 958.22 |
| 159351-69-6 | |
| C53H83NO14 | |
| 10 mg | |
| Soluble in dimethysulfoxide,ethanol and chloroform; Slightly soluble in water. | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC | |
| 129317877 | |
| Powder |
Chemical Identifiers
| 159351-69-6 | |
| 958.24 | |
| HKVAMNSJSFKALM-PWMUVMERSA-N | |
| CC1CCC2CC(C(=CC=CC=CC(CC(C(=O)C(C(C(=CC(C(=O)CC(OC(=O)C3CCCCN3C(=O)C(=O)C1(O2)O)C(C)CC4CCC(C(C4)OC)OCCO)C)C)O)OC)C)C)C)OC |
| C53H83NO14 | |
| MFCD07785165 | |
| 129317877 |
Safety and Handling
GHS H Statement
H372
Causes damage to organs through prolonged or repeated exposure.
P260-P264b-P270-P314-P501c
H372
TSCA : No
Recommended Storage : Store at -20°C
RUO – Research Use Only