missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Caplugs™ Evergreen Scientific Two-Position Stopper Caps
For use with 17mm polypropylene culture tubes
Supplier: Caplugs™ 300 3817 02P
Description
- Over-fitting caps have two positions: loose for aerobic culturing, or tight for anaerobic culturing and leakproof sealing
- Gamma-irradiated for sterility
- Color: Natural
Specifications
| 2-position Stopper Cap | |
| Natural | |
| 17 mm | |
| 17 mm Tubes |
| Polypropylene | |
| 0.67 in. | |
| Sterile |