missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Expert pH Pocket Tester

Catalog No. 13643133
Change view
Click to view available options
Measurement Modes:
pH, Temperature
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Modes
13643133 pH, Temperature
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13643133 Supplier Thermo Scientific™ Supplier No. EXPERTPH

Perform on-the-go, fuss-free measurements for applications such as water treatment, environmental monitoring, education, hydroponics, agriculture, aquaculture, food & beverage and many more!

The Thermo Scientific™ Expert pH Pocket Tester is designed for on-the-go measurements of pH and temperature. The Expert pH Pocket Tester is best used in applications such as hydroponic gardening, aquaculture, agriculture, pools, cooling towers, and more!

  • Large 4 digit double line display
  • Simultaneous display of pH and temperature
  • Battery life, ready, and calibration indicators
  • Automatic temperature compensation
  • Auto buffer recognition and calibration
  • Multi language operational instructions

Display:

  • Large 4 digit double line display and wide viewing angle make it easy to read and record measurements
  • Battery life, ready, and calibration indicators
  • Simultaneous display of pH with temperature

Keypad:

  • Easy to operate and intuitive user interface with power, calibration, menu, and enter keys
  • One push calibration: immerse sensor in calibration solution and press Cal
  • Auto pH buffer recognition and calibration

Cap:

  • Gasketed and leak proof with an O-ring seal
  • Transparent cap with a fill line indicator
  • Symmetrical cap can be front or rear facing
  • Cap secures to the top of tester when not in use

Additional features:

  • Lanyard provision enables the user to easily transport the pocket tester for on-the-go measurements
  • Pocket tester floats and meets Waterproof IP67 rating
  • Wide, stable base allows the tester to independently stand vertically for measurements
  • Automatic temperature compensation: adjusts readings according to temperature change for greater accuracy
  • Auto-off function: pocket tester turns off after 8.5 minutes of non-use to extend battery life
  • CD/RoHS Marking
  • Multi language instructions available

Specifications

Accuracy ±0.1 pH
Accuracy (pH) ±0.1
Backlight Option No
Battery Life 100 hr. (average)
Battery Type 4 x 1.5V Micro Alkaline
Calibration pH, temperature
Calibration Points 1 to 3
Certifications/Compliance CE, FCC Class A, ROHS-2
Description Expert pH pocket tester for measuring pH with temperature
Display Type 4-digit, Double-line
IP Rating IP67
Measurement Modes pH, Temperature
Range (pH) 0 to 14
Resolution (pH) 0.1
Temperature Range (Metric) 0.0°C to 60.0°C
Includes Expert pH tester with batteries
Relative Humidity Range 5 to 85%, non-condensing
Resolution 0.1pH
Temperature Accuracy ±0.6°C, ±1.0°F
Temperature Compensation Yes
Temperature (English) Operating 32.0°F to 122.0°F
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature Probe Calibration 1 point
Temperature Range (English) 32.0°F to 140.0°F
Temperature Resolution 0.1°C, 0.1°F
Warranty 1 Year
Length (English) 6.7 in.
Length (Metric) 17 cm
Width (English) 1.8 in.
Width (Metric) 4.5 cm
Height (English) 1.2 in.
Height (Metric) 3 cm
Weight (English) 3.4 oz.
Weight (Metric) 96.3 g
Show More Show Less