missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Expert CTS Pocket Tester

Catalog No. 13643132
Change view
Click to view available options
Measurement Modes:
Conductivity, TDS, Salinity, Temperature
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Measurement Modes
13643132 Conductivity, TDS, Salinity, Temperature
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 13643132 Supplier Thermo Scientific™ Supplier No. EXPERTCTS

Ensure accurate measurements of Conductivity, TDS, Salinity, and Temperature using the easy-to-use, large 4-digit display pocket tester for many applications.

The Thermo Scientific™ Expert CTS Pocket Tester is designed for on-the-go measurements such as – Conductivity, TDS, Salinity, Temperature and other related electrochemistry measurements. The Expert CTS Pocket Tester is best used in applications such as environmental monitoring, electroplating, agriculture, education and more!

  • Large 4 digit double line display
  • Battery life, ready, and calibration indicators
  • Automatic temperature compensation
  • Auto buffer recognition and calibration
  • Multi language operational instructions

Display:

  • Large 4 digit double line display and wide viewing angle make it easy to read and record measurements
  • Battery life, ready, and calibration indicators
  • Simultaneous display of conductivity/TDS/salinity with temperature

Keypad:

  • Easy to operate and intuitive user interface with power, calibration, menu, and enter keys
  • One push calibration: immerse sensor in calibration solution and press Cal
  • Auto buffer recognition and calibration
  • Cap: Gasketed and leak proof with an O-ring seal
  • Transparent cap with a fill line indicator
  • Symmetrical cap can be front or rear facing
  • Cap secures to the top of tester when not in use

Additional features:

  • Lanyard provision enables the user to easily transport the pocket tester for on-the-go measurements
  • Pocket tester floats and meets Waterproof IP67 rating
  • Wide, stable base allows the tester to independently stand vertically for measurements
  • Automatic temperature compensation: adjusts readings according to temperature change for greater accuracy
  • Auto-off function: pocket tester turns off after 8.5 minutes of non-use to extend battery life
  • CE/RoHS Marking
  • Multi language instructions available

Specifications

Accuracy ±2% full scale ±1LSD
Accuracy (Conductivity) ±2% full scale
Accuracy (Salinity) ±2% full scale
Accuracy (TDS) ±2% full scale
Backlight Option No
Battery Life 75 hr. (average)
Battery Type 4 x 1.5V Micro Alkaline
Calibration Auto and Manual Calibration
Calibration Modes Conductivity, TDS, salinity, temperature
Calibration Points 1
Calibration Points (Conductivity) 1
Certifications/Compliance CE, FCC Class A, ROHS-2
Compensation Automatic Temperature Compensation
Description Expert CTS pocket tester for measuring conductivity, TDS or salinity with temperature
Display Type 4-digit, Double-line
IP Rating IP67
Measurement Modes Conductivity, TDS, Salinity, Temperature
Range (Conductivity) 0.00 to 20.00 mS/cm
Range (Salinity) 0.0 to 10.0 ppt
Range (TDS) 0.00 to 20.00 ppt
Temperature Range (Metric) 0°C to 50.0°C
Includes Expert CTS tester with batteries
Product Type Pocket tester
Range Conductivity: 0.00 to 20.00mS; TDS: 0.00 to 20.00 ppt (with TDS Factor 1.00); Salinity: 0.0 to 10.0 ppt
Relative Humidity Range 5 to 85 %, non-condensing
Resolution 电导率:10uS <2.00mS, 0.10mS >2.00mS; TDS: 10 ppm <2.00ppt, 0.10 ppt >2.00ppt; 盐度:0.10 ppt
Resolution (Conductivity) 10 μS/cm for 0 to 1990 μS/cm; 0.10 mS/cm for 2.00 to 20.00 mS/cm
Resolution (Salinity) 0.1 ppt
Resolution (TDS) 10 ppm for 0 to 1990 ppm; 0.10 ppt for 2.00 to 20.00 ppt
Salinity Measurement Modes Per practical salinity curve
TDS Factor Range 0.40 to 1.00 (selectable)
Temperature Accuracy ±0.6°C, ±1.0°F
Temperature Compensation Yes
Temperature (English) Operating 32°F to 122°F
Temperature (Metric) Operating 5°C to 45°C (ambient)
Temperature Probe Calibration 1 point
Temperature Range (English) 32.0°F to 122.0°F
Temperature Resolution 0.1°C, 0.1°F
Warranty 1 Year
Length (English) 6.7 in.
Length (Metric) 17 cm
Width (English) 1.8 in.
Width (Metric) 4.5 cm
Height (English) 1.2 in.
Height (Metric) 3 cm
Weight (English) 3.3 oz.
Weight (Metric) 93.7 g
Show More Show Less