missing translation for 'onlineSavingsMsg'
Learn More

Europium, plasma standard solution, Specpure™ Eu 10,000μg/mL

$233.01 - $451.92

Chemical Identifiers

MDL Number MFCD00010992
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity  
Catalog Number Mfr. No. Quantity Price Quantity  
AA35761AE
View Documents
Thermo Scientific Chemicals
035761AE
100 mL N/A
Only null left
 
AA35761AP
View Documents
Thermo Scientific Chemicals
035761AP
500 mL N/A
Only null left
 
Description

Description

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Specifications

Chemical Identifiers

MFCD00010992

Specifications

Liquid
MFCD00010992
100 mL
Europium plasma standard solution, Eu 10000μg/mL
Eu2O3 in 5% HNO3
UN3264
Specpure
Videos
Safety and Handling

Safety and Handling

GHS H Statement
H314
Causes severe skin burns and eye damage.

P234-P260-P264b-P271-P280-P301+P330+P331-P303+P361+P353-P304+P340-P305+P351+P338-P310-P363-P390-P501c

H290-H314-H335

DOTInformation : Transport Hazard Class: 8; Packing Group: III; Proper Shipping Name: CORROSIVE LIQUID, ACIDIC, INORGANIC, N.O.S.

EINECSNumber : 231-714-2

TSCA : Yes

Recommended Storage : Ambient temperatures

SDS
Documents

Documents

RUO – Research Use Only