Learn More
Europium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific™
$766.14 - $766.14
Chemical Identifiers
| CAS | 13537-22-9 |
|---|---|
| Molecular Formula | Cl3EuO12 |
| Molecular Weight (g/mol) | 450.30 |
| MDL Number | MFCD00016076 |
| InChI Key | KDBBDPJZIDRZFY-UHFFFAOYSA-K |
| Synonym | europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol. |
| PubChem CID | 11733073 |
| IUPAC Name | europium(3+);triperchlorate |
| SMILES | [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
Description
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Chemical Identifiers
| 13537-22-9 | |
| 450.30 | |
| KDBBDPJZIDRZFY-UHFFFAOYSA-K | |
| 11733073 | |
| [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O |
| Cl3EuO12 | |
| MFCD00016076 | |
| europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol. | |
| europium(3+);triperchlorate |
Specifications
| 13537-22-9 | |
| 10 g | |
| MFCD00016076 | |
| europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol. | |
| KDBBDPJZIDRZFY-UHFFFAOYSA-K | |
| europium(3+);triperchlorate | |
| 11733073 | |
| Reagent | |
| Odorless |
| Liquid | |
| Cl3EuO12 | |
| UN3211 | |
| Insoluble in water. | |
| [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O | |
| 450.30 | |
| 558.40 (450.32 Anhydrous) | |
| 50% w/w aq. soln. | |
| Europium(III) perchlorate hexahydrate |
Safety and Handling
GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
H271-H315-H319-H335
DOTInformation : Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber : 236-907-5
TSCA : No
Recommended Storage : Ambient temperatures