accessibility menu, dialog, popup

UserName

CAS RN 13537-22-9

IUPAC Name: europium(3+) triperchlorate
Synonyms: europium(3+) triperchlorate
Molecular Weight (g/mol): 450.3
Molecular Formula: Cl3EuO12
InChi Key: KDBBDPJZIDRZFY-UHFFFAOYSA-K
SMILES: [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results