missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) perchlorate hexahydrate, 50% w/w in aq. soln., Reagent Grade, Thermo Scientific™

Catalog No. AA40572NC
Change view
Click to view available options
Quantity:
10 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA40572NC 10 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. AA40572NC Supplier Thermo Scientific Chemicals Supplier No. 40572NC

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Chemical Identifiers

CAS 13537-22-9
Molecular Formula Cl3EuO12
Molecular Weight (g/mol) 450.30
MDL Number MFCD00016076
InChI Key KDBBDPJZIDRZFY-UHFFFAOYSA-K
Synonym europium perchlorate, europium 3+ perchlorate, acmc-1aedp, europium iii perchlorate solution, trihyperchloric acid europium iii salt, europium iii perchlorate aqueous sol.
PubChem CID 11733073
IUPAC Name europium(3+);triperchlorate
SMILES [Eu+3].[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O.[O-][Cl](=O)(=O)=O

Specifications

Physical Form Liquid
Quantity 10 g
UN Number UN3211
Solubility Information Insoluble in water.
Formula Weight 558.40 (450.32 Anhydrous)
Grade Reagent
Concentration or Composition (by Analyte or Components) 50% w/w aq. soln.
Odor Odorless
Chemical Name or Material Europium(III) perchlorate hexahydrate
Hazard Category H271-H315-H319-H335
Hazard Statement GHS H Statement
H272-H315-H319-H335
May intensify fire
oxidizer.
Causes skin irritation.
Causes serious eye irritation.
May cause respiratory irritation.
Precautionary Statement P210-P220-P221-P261-P264b-P271-P280-P302+P352-P304+P340-P305+P351+P338-P312-P332+P313-P362-P370+P378q-P501c
DOTInformation Transport Hazard Class: 5.1; Packing Group: II; Proper Shipping Name: PERCHLORATES, INORGANIC, AQUEOUS SOLUTION, N.O.S.
EINECSNumber 236-907-5
TSCA No
Recommended Storage Ambient temperatures