missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, REacton™, 99.996% (REO)

Catalog No. p-7059294
Change view
missing translation for 'orderingAttributeHoverText'
Quantity:
100 g
25 g
5 g
Catalog No. Quantity
AA4434706 5 g
AA4434714 25 g
AA4434722 100 g
3 options

Catalog No. AA4434706

Supplier: Thermo Scientific Chemicals 04434706

Only null left

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes, red or blue phosphor in television sets. It shows an useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
IUPAC Name dieuropium(3+) trioxidandiide
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Physical Form Powder
Quantity 5 g
Merck Index 14,3904
Solubility Information Insoluble in water.
Formula Weight 351.92
Percent Purity 99.996%
Grade Reacton
Sensitivity Hygroscopic
Assay (REO)
Chemical Name or Material Europium (III) oxide
Show More Show Less
EINECSNumber 215-165-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only