accessibility menu, dialog, popup

UserName

CAS RN 1308-96-9

IUPAC Name: dieuropium(3+) trioxidandiide
Synonyms: europium oxide
Molecular Weight (g/mol): 351.93
Molecular Formula: Eu2O3
InChi Key: AEBZCFFCDTZXHP-UHFFFAOYSA-N
SMILES: [O--].[O--].[O--].[Eu+3].[Eu+3]
Molecular Weight (g/mol) 
  • (2)
  • (17)
Physical Form 
  • (14)
  • (1)
Quantity 
  • (2)
  • (3)
  • (1)
  • (5)
  • (4)
Percent Purity 
  • (3)
  • (2)
  • (3)
Grade 
  • (1)
  • (3)
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (2)
  • (2)
  • (15)
  • (2)
  • (17)
  • (14)
  • (1)
  • (2)
  • (3)
  • (1)
  • (5)
  • (4)
  • (3)
  • (2)
  • (3)
  • (1)
  • (3)
  • (2)
  • (1)
  • (1)