missing translation for 'onlineSavingsMsg'
Learn More

Europium(III) oxide, REacton™, 99.9% (REO)

Catalog No. p-4408089
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AA1130006 5 g
AA1130014 25 g
AA1130022 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AA1130006 Supplier Thermo Scientific Chemicals Supplier No. 01130006

CAS: 1308-96-9 | Eu2O3 | 351.93 g/mol

Europium(III) oxide is used as an agent for the manufacture of fluorescent glass. It can be used in the anti-counterfeiting phosphors in euro banknotes and red or blue phosphor in television sets. It shows a useful application as an activator for yttrium based phosphors.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 1308-96-9
Molecular Formula Eu2O3
Molecular Weight (g/mol) 351.93
MDL Number MFCD00010997
InChI Key AEBZCFFCDTZXHP-UHFFFAOYSA-N
Synonym europium oxide, europia, europium trioxide, dieuropium trioxide, europium sesquioxide, europium 3+ oxide, europium oxide euo1.5, europium 3+ ; oxygen 2-
PubChem CID 159371
IUPAC Name europium(3+);oxygen(2-)
SMILES [O--].[O--].[O--].[Eu+3].[Eu+3]

Specifications

Melting Point 2291°C
Color White
Physical Form Powder
Quantity 5 g
Assay Percent Range 99.9% (REO)
Solubility Information Insoluble in water.
Formula Weight 351.92
Sensitivity Hygroscopic
Appearance White powder or chunks
Density 7.42g/cm3
Chemical Name or Material Europium(III) oxide
Show More Show Less
EINECSNumber 215-165-6
TSCA Yes
Recommended Storage Ambient temperatures

RUO – Research Use Only