missing translation for 'onlineSavingsMsg'
Learn More

Eugenol, USP, Spectrum™ Chemical

Catalog No. 18602596
Change view
Click to view available options
Quantity:
100 mL
500 mL
4 L
20 L
200 L
Packaging:
Amber Glass Bottle
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
18602596 200 L Amber Glass Bottle
18602592 100 mL Amber Glass Bottle
18602593 500 mL Amber Glass Bottle
18602594 4 L Amber Glass Bottle
18602595 20 L Amber Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. 18602596 Supplier Spectrum Chemical Mfg Cor Supplier No. EU110200LTBL

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

EU110, 97-53-0, C10H12O2

Spectrum™ Chemical Eugenol, USP is used as a local antiseptic and anaesthetic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
IUPAC Name 2-methoxy-4-(prop-2-en-1-yl)phenol
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

CAS Max % 100%
Boiling Point 253°C
Packaging Amber Glass Bottle
Refractive Index 1.540-1.542
Quantity 200 L
Grade USP
Physical Form Liquid