missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Eugenol, USP, Spectrum™ Chemical
EU110, 97-53-0, C10H12O2
Supplier: Spectrum Chemical Mfg Cor EU11020LTBL
Description
Spectrum™ Chemical Eugenol, USP is used as a local antiseptic and anaesthetic. All Spectrum Chemical USP grade products are manufactured, packaged and stored under current Good Manufacturing Practices (cGMP) per 21CFR part 211 in FDA registered and inspected facilities.Specifications
| 97-53-0 | |
| 253°C | |
| C10H12O2 | |
| MFCD00008654 | |
| RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
| 2-methoxy-4-(prop-2-en-1-yl)phenol | |
| USP |
| 100% | |
| Amber Glass Bottle | |
| 1.540-1.542 | |
| 20 L | |
| COC1=CC(CC=C)=CC=C1O | |
| 164.20 | |
| Liquid |
Chemical Identifiers
| 97-53-0 | |
| 164.20 | |
| RRAFCDWBNXTKKO-UHFFFAOYSA-N | |
| COC1=CC(CC=C)=CC=C1O |
| C10H12O2 | |
| MFCD00008654 | |
| 2-methoxy-4-(prop-2-en-1-yl)phenol |