missing translation for 'onlineSavingsMsg'
Learn More

Eugenol 99.0+%, TCI America™

Catalog No. A023225ML
Change view
Click to view available options
Quantity:
25 mL
100 mL
500 mL
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
A023225ML 25 mL
A0232100ML 100 mL
A0232500ML 500 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. A023225ML Supplier TCI America Supplier No. A023225ML

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
Synonym eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
PubChem CID 3314
ChEBI CHEBI:4917
IUPAC Name 2-methoxy-4-(prop-2-en-1-yl)phenol
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

Melting Point -9°C
Color Yellow
Boiling Point 254°C
Quantity 25 mL
Formula Weight 164.20
Percent Purity ≥99.0% (GC)
Physical Form Liquid
Chemical Name or Material Eugenol
EINECSNumber (3)-0637
RTECSNumber SJ4375000
TSCA Yes