missing translation for 'onlineSavingsMsg'
Learn More

Eugenol, 99%

Catalog No. p-7021835
Change view
Click to view available options
Quantity:
100 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AAA1433222 100 g
AAA1433236 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AAA1433222 Supplier Thermo Scientific Chemicals Supplier No. A1433222
Only null left

CAS: 97-53-0 | C10H12O2 | 164.20 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Suitable for battery materials development.

Applications
Internal standard in GC assays for presence of thymol in biological samplesEugenol is used in cosmetics, essential oil, perfumeries and flavorings. It finds application as a local antiseptic and anesthetic. It is combined with zinc oxide to form zinc oxide eugenol, which is used in dental restoration especially for root canal sealing and prosthodontics. It is also an antifungal antimicrobial agonist of the TRPV1 receptor. It is also used in production of stabilizers and antioxidants for plastics and rubbers. Further, it is used in the production of vanillin. It serves as an active ingredient in cloves. In addition to this, it is used as an insect attractant, chemical intermediate and denaturant for alcohol.

Solubility
Miscible with ethanol, ether, oils, alcohol, glacial acetic acid, chloroform and aqueous sodium hydroxide. Immiscible with water.

Notes
Air sensitive. Incompatible with strong oxidizing agents.

Chemical Identifiers

CAS 97-53-0
Molecular Formula C10H12O2
Molecular Weight (g/mol) 164.20
MDL Number MFCD00008654
InChI Key RRAFCDWBNXTKKO-UHFFFAOYSA-N
Synonym eugenol, 4-allylguaiacol, 4-allyl-2-methoxyphenol, eugenic acid, allylguaiacol, caryophyllic acid, p-allylguaiacol, p-eugenol, 2-methoxy-4-allylphenol, engenol
PubChem CID 3314
ChEBI CHEBI:4917
IUPAC Name 2-methoxy-4-prop-2-enylphenol
SMILES COC1=CC(CC=C)=CC=C1O

Specifications

Melting Point -10°C
Density 1.068
Boiling Point 252°C to 253°C
Flash Point 127°C (260°F)
Odor Pungent
Refractive Index 1.541
Quantity 100 g
Beilstein 1366759
Sensitivity Air Sensitive
Merck Index 14,3898
Solubility Information Miscible with ethanol,ether,oils,alcohol,glacial acetic acid,chloroform and aqueous sodium hydroxide. Immiscible with water.
Formula Weight 164.21
Percent Purity 99%
Chemical Name or Material Eugenol
Show More Show Less
Hazard Category H302-H315-H317-H319
Hazard Statement GHS H Statement
H302-H315-H319-H317
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
May cause an allergic skin reaction.
Precautionary Statement P261-P264b-P270-P272-P280-P301+P312-P302+P352-P305+P351+P338-P330-P333+P313-P362-P501c
EINECSNumber 202-589-1
RTECSNumber SJ4375000
TSCA Yes
Recommended Storage Ambient temperatures; Store under Argon

RUO – Research Use Only