missing translation for 'onlineSavingsMsg'
Learn More

Etomidate 98.0+%, TCI America™

Catalog No. p-7129209
Change view
Click to view available options
Quantity:
100 mg
500 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E0897100MG 100 mg
E0897500MG 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E0897100MG Supplier TCI America Supplier No. E0897100MG
Only null left

Chemical Identifiers

CAS 33125-97-2
Molecular Formula C14H16N2O2
Molecular Weight (g/mol) 244.294
MDL Number MFCD00869295
InChI Key NPUKDXXFDDZOKR-NSHDSACASA-N
Synonym etomidate, +-etomidate, d-etomidate, etomidato, etomidatum, amidate, hypnomidate, r-ethyl 1-1-phenylethyl-1h-imidazole-5-carboxylate, etomidatum inn-latin
PubChem CID 6338045
IUPAC Name ethyl 3-[(1S)-1-phenylethyl]imidazole-4-carboxylate
SMILES CCOC(=O)C1=CN=CN1C(C)C2=CC=CC=C2

Specifications

Melting Point 69°C
Color White
Quantity 100 mg
UN Number 3077
Formula Weight 244.29
Percent Purity ≥98.0% (GC)
Physical Form Crystalline Powder
Chemical Name or Material Etomidate
RTECSNumber NI4021500
TSCA No
Recommended Storage Refrigerator