CAS RN 33125-97-2
CAS RN 33125-97-2
IUPAC Name:
ethyl 1-(1-phenylethyl)-1H-imidazole-5-carboxylate
Synonyms:
etomidate etomidate (substance) ethomidate d-etomidate etomidate (product) (d)-etomidate D-etomidate amidate (pharmaceutical) radenarcon radenarkon
Molecular Weight (g/mol):
244.29
Molecular Formula:
C14H16N2O2
InChi Key:
NPUKDXXFDDZOKR-UHFFFAOYNA-N
SMILES:
CCOC(=O)C1=CN=CN1C(C)C1=CC=CC=C1
1
–
1
of
1
results
1
Etomidate 98.0+%, TCI America™
CAS: 33125-97-2 Molecular Formula: C14H16N2O2 Molecular Weight (g/mol): 244.294 MDL Number: MFCD00869295 InChI Key: NPUKDXXFDDZOKR-NSHDSACASA-N Synonym: etomidate,+-etomidate,d-etomidate,etomidato,etomidatum,amidate,hypnomidate,r-ethyl 1-1-phenylethyl-1h-imidazole-5-carboxylate,etomidatum inn-latin PubChem CID: 6338045 IUPAC Name: ethyl 3-[(1S)-1-phenylethyl]imidazole-4-carboxylate SMILES: CCOC(=O)C1=CN=CN1C(C)C2=CC=CC=C2