Learn More
Ethynyltri-n-butyltin, 96%
CAS: 994-89-8 | C14H28Sn | 315.088 g/mol
$139.68 - $452.03
Chemical Identifiers
| CAS | 994-89-8 |
|---|---|
| Molecular Formula | C14H28Sn |
| Molecular Weight (g/mol) | 315.088 |
| MDL Number | MFCD00009420 |
| InChI Key | YEMJHNYABQHWHL-UHFFFAOYSA-N |
| Synonym | tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin |
| PubChem CID | 621176 |
| IUPAC Name | tributyl(ethynyl)stannane |
| SMILES | CCCC[Sn](CCCC)(CCCC)C#C |
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Price | Quantity | |||||
|
AAL2017803
|
Thermo Scientific Chemicals
L2017803 |
1 g |
Each for $139.68
|
|
|||||
|
AAL2017806
|
Thermo Scientific Chemicals
L2017806 |
5 g |
Each for $452.03
|
|
|||||
Description
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
ApplicationsUsed to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.
Solubility
Not miscible or difficult to mix with water.
Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
Chemical Identifiers
| 994-89-8 | |
| 315.088 | |
| YEMJHNYABQHWHL-UHFFFAOYSA-N | |
| 621176 | |
| CCCC[Sn](CCCC)(CCCC)C#C |
| C14H28Sn | |
| MFCD00009420 | |
| tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin | |
| tributyl(ethynyl)stannane |
Specifications
| 994-89-8 | |
| 70°C (0.2 mmHg) | |
| C14H28Sn | |
| HC≡C-Sn(CH2CH2CH2CH3)3 | |
| 1 g | |
| 3537659 | |
| tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin | |
| YEMJHNYABQHWHL-UHFFFAOYSA-N | |
| tributyl(ethynyl)stannane | |
| 621176 | |
| 96% |
| 1.08 | |
| 73°C (163°F) | |
| 1.477 | |
| MFCD00009420 | |
| UN2788 | |
| Moisture sensitive | |
| Not miscible or difficult to mix with water. | |
| CCCC[Sn](CCCC)(CCCC)C#C | |
| 315.088 | |
| 315.09 | |
| Ethynyltri-n-butyltin |
Safety and Handling
GHS H Statement
H301-H372-H227-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.
P201-P202-P210-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P501c
H227-H301-H312-H315-H319-H360FD-H372
DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.
TSCA : No
Recommended Storage : Keep cold
RUO – Research Use Only