missing translation for 'onlineSavingsMsg'
Learn More

Ethynyltri-n-butyltin, 96%

CAS: 994-89-8 | C14H28Sn | 315.088 g/mol

Supplier:  Thermo Scientific Chemicals L2017806

 View more versions of this product

Catalog No. AAL2017806


Only null left
Explore available promotions

Description

Description

Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Used to prepare vinylstannanes and stannylisoxazoles. Ethynylation reagent for azaaromatics.

Solubility
Not miscible or difficult to mix with water.

Notes
Mositure sensitive. Store away from oxidizing agents and water/moisture.
Specifications

Specifications

Ethynyltri-n-butyltin
1.08
73°C (163°F)
1.477
MFCD00009420
UN2788
Moisture sensitive
Not miscible or difficult to mix with water.
CCCC[Sn](CCCC)(CCCC)C#C
315.088
315.09
994-89-8
70°C (0.2 mmHg)
C14H28Sn
HC≡C-Sn(CH2CH2CH2CH3)3
5 g
3537659
tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
YEMJHNYABQHWHL-UHFFFAOYSA-N
tributyl(ethynyl)stannane
621176
96%

Chemical Identifiers

994-89-8
315.088
YEMJHNYABQHWHL-UHFFFAOYSA-N
621176
CCCC[Sn](CCCC)(CCCC)C#C
C14H28Sn
MFCD00009420
tributylstannylacetylene, ethynyltri-n-butyltin, ethynyltributylstannane, tributyl ethynyl stannane, stannane, tributylethynyl, tributylethynyl tin, ethynyltributyltin, tributylethynyltin, tributylstannylethyne, tributyl ethynyl tin
tributyl(ethynyl)stannane
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

GHS H Statement
H301-H372-H227-H312-H315-H319
Toxic if swallowed.
Causes damage to organs through prolonged or repeated exposure.
Combustible liquid.
Harmful in contact with skin.
Causes skin irritation.
Causes serious eye irritation.

P201-P202-P210-P260-P264b-P270-P280i-P281-P301+P310-P302+P352-P305+P351+P338-P308+P313-P312-P330-P332+P313-P362-P370+P378q-P501c

H227-H301-H312-H315-H319-H360FD-H372

DOTInformation : Transport Hazard Class: 6.1; Packing Group: III; Proper Shipping Name: ORGANOTIN COMPOUNDS, LIQUID, N.O.S.

TSCA : No

Recommended Storage : Keep cold

SDS
Documents

Documents

Promotions

Promotions

RUO – Research Use Only