Learn More
Ethynylmagnesium chloride, 0.5M solution in THF/Toluene, AcroSeal™
CAS: 65032-27-1 | C2HClMg | 84.79 g/mol
$100.35 - $539.22
Chemical Identifiers
| CAS | 65032-27-1 |
|---|---|
| Molecular Formula | C2HClMg |
| Molecular Weight (g/mol) | 84.79 |
| MDL Number | MFCD00075343 |
| InChI Key | YEWNLGTVYUADOR-UHFFFAOYSA-M |
| Synonym | ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf |
| PubChem CID | 2734899 |
| IUPAC Name | chloro(ethynyl)magnesium |
| SMILES | Cl[Mg]C#C |
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|---|---|---|---|---|---|---|---|---|---|
| Catalog Number | Mfr. No. | Quantity | Packaging | Price | Quantity | ||||
|
AC388951000
|
Thermo Scientific Chemicals
388951000 |
100 mL | AcroSeal™ Glass bottle |
Each for $100.35
|
|
||||
|
AC388958000
|
Thermo Scientific Chemicals
388958000 |
800 mL | AcroSeal™ Glass Bottle |
Each for $539.22
|
|
||||
Description
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Chemical Identifiers
| 65032-27-1 | |
| 84.79 | |
| YEWNLGTVYUADOR-UHFFFAOYSA-M | |
| 2734899 | |
| Cl[Mg]C#C |
| C2HClMg | |
| MFCD00075343 | |
| ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf | |
| chloro(ethynyl)magnesium |
Specifications
| Brown | |
| C2HClMg | |
| MFCD00075343 | |
| 0.9600g/mL | |
| Solubility in water: reacts. | |
| Cl[Mg]C#C | |
| 84.79 | |
| 84.79 | |
| 0.96 | |
| Liquid |
| 65032-27-1,108-88-3,109-99-9 | |
| HC≡CMgCl | |
| 4 to 6% | |
| ethynylmagnesium chloride, ethynylmagnesiumchloride, ethynylchloromagnesium, ethynyl magnesium chloride, ethynyl-magnesium chloride, chloro ethynyl magnesium, yewnlgtvyuador-uhfffaoysa-m, ethynylmagnesium chloride solution, 0.5 m in thf | |
| YEWNLGTVYUADOR-UHFFFAOYSA-M | |
| chloro(ethynyl)magnesium | |
| 2734899 | |
| AcroSeal™ Glass bottle | |
| 100 mL | |
| Ethynylmagnesium chloride, 0.6M solution in THF/Toluene |
Safety and Handling
GHS H Statement
Highly flammable liquid and vapor. In contact with water releases flammable gases which may ignite spontaneously. Causes severe skin burns and eye damage. May cause respiratory irritation. Suspected of damaging the unborn child. Reacts violently with water. May form explosive peroxides. Suspected of causing cancer. May be fatal if swallowed and enters airways. May cause damage to organs through prolonged or repeated exposure if inhaled. May cause drowsiness or dizziness.
GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces. - No smoking. IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/shower. Handle under inert gas. Protect from moisture. Wear protective gloves/protective clothing/eye protection/face protection. IF SWALLOWED: rinse mouth. Do NOT induce vomiting. IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing. Immediately call a POISON CENTER or doctor/physician.
Danger
Recommended Storage : Product tends to crystallize at lower temperatures. Store above 15°C. Precipitates can be redissolved by warming and appropriate homogenization.
RUO – Research Use Only