missing translation for 'onlineSavingsMsg'
Learn More

Ethynylmagnesium bromide, 0.5M solution in THF, AcroSeal™

Catalog No. p-4229006
Change view
Click to view available options
Quantity:
100 mL
800 mL
Packaging:
AcroSeal™ Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC439121000 100 mL AcroSeal™ Glass Bottle
AC439128000 800 mL AcroSeal™ Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. AC439121000 Supplier Thermo Scientific Chemicals Supplier No. 439121000

CAS: 4301-14-8 | C2HBrMg | 129.24 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Chemical Identifiers

CAS 4301-14-8 , 109-99-9
Molecular Formula C2HBrMg
Molecular Weight (g/mol) 129.24
MDL Number MFCD00075342
InChI Key HUGJUYPSXULVQQ-UHFFFAOYSA-M
Synonym ethynylmagnesium bromide, bromo ethynyl magnesium, ethynylmagnesium bromide solution, ethinylmagnesium bromide, ethynyl magnesiumbromide, pubchem18180, ethynyl-magnesium bromide, ethinyl magnesium bromide, ethynyl magnesium bromide, hugjuypsxulvqq-uhfffaoysa-m, grignard reagent
PubChem CID 4071243
IUPAC Name bromo(ethynyl)magnesium
SMILES Br[Mg]C#C

Specifications

Color Colorless to Gray
Physical Form Liquid
Quantity 100 mL
Linear Formula CHCMgBr
Flash Point −17°C
Formula Weight 129.24
Concentration or Composition (by Analyte or Components) 0.40 to 0.60M
Packaging AcroSeal™ Glass Bottle
Specific Gravity 0.94
Density 0.9400g/mL
Chemical Name or Material Ethynylmagnesium bromide
Show More Show Less
Hazard Category GHS Signal Word: Danger
Hazard Statement GHS H Statement
Highly flammable liquid and vapor.
In contact with water releases flammable gases which may ignite spontaneously.
Causes severe skin burns and eye damage.
May cause respiratory irritation.
Reacts violently w
Precautionary Statement GHS P Statement
Keep away from heat/sparks/open flames/hot surfaces.
- No smoking.
IF ON SKIN (or hair): Take off immediately all contaminated clothing.
Rinse skin with water/shower.
Handle under inert gas.
Protect from moisture

RUO – Research Use Only