missing translation for 'onlineSavingsMsg'
Learn More

Ethynylbenzene 98.0+%, TCI America™

Supplier:  TCI America E0196100ML

 View more versions of this product

Catalog No. E0196100ML


Only null left
Explore available promotions

Description

Description

Specifications

Specifications

Ethynylbenzene
Yellow
C8H6
100 mL
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
C#CC1=CC=CC=C1
102.136
102.14
Liquid
536-74-3
142°C
MFCD00008570
3295
UEXCJVNBTNXOEH-UHFFFAOYSA-N
ethynylbenzene
10821
≥98.0% (GC)

Chemical Identifiers

536-74-3
102.136
UEXCJVNBTNXOEH-UHFFFAOYSA-N
10821
C#CC1=CC=CC=C1
C8H6
MFCD00008570
phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
ethynylbenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

RTECSNumber : DA0780000

TSCA : Yes

Recommended Storage : Refrigerator

SDS
Documents

Documents

Promotions

Promotions