missing translation for 'onlineSavingsMsg'
Learn More

Ethynylbenzene 98.0+%, TCI America™

Catalog No. p-7128892
Change view
Click to view available options
Quantity:
25 mL
100 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019625ML 25 mL
E0196100ML 100 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E019625ML Supplier TCI America Supplier No. E019625ML
Only null left

Chemical Identifiers

CAS 536-74-3
Molecular Formula C8H6
Molecular Weight (g/mol) 102.136
MDL Number MFCD00008570
InChI Key UEXCJVNBTNXOEH-UHFFFAOYSA-N
Synonym phenylacetylene, phenylethyne, benzene, ethynyl, 1-phenylethyne, phenylacetylide, phenyl acetylene, ethyne, phenyl, acetylene, phenyl, ethynyl-benzene, phenylacethylene
PubChem CID 10821
IUPAC Name ethynylbenzene
SMILES C#CC1=CC=CC=C1

Specifications

Color Yellow
Boiling Point 142°C
Quantity 25 mL
UN Number 3295
Formula Weight 102.14
Percent Purity ≥98.0% (GC)
Physical Form Liquid
Chemical Name or Material Ethynylbenzene
RTECSNumber DA0780000
TSCA Yes
Recommended Storage Refrigerator