missing translation for 'onlineSavingsMsg'
Learn More

Ethynyl(phenyl)iodonium Tetrafluoroborate 97.0+%, TCI America™

Catalog No. E04671G
Change view
Click to view available options
Quantity:
1 g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E04671G 1 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. E04671G Supplier TCI America Supplier No. E04671G

Please call Customer Service at 1-800-234-7437 or send an email to help@thermofisher.com for assistance.

Chemical Identifiers

CAS 127783-34-0
Molecular Formula C8H6BF4I
Molecular Weight (g/mol) 315.844
MDL Number MFCD00191648
InChI Key AVSBPVBKCONUGF-UHFFFAOYSA-N
PubChem CID 2758943
IUPAC Name ethynyl(phenyl)iodanium;tetrafluoroborate
SMILES [B-](F)(F)(F)F.C#C[I+]C1=CC=CC=C1

Specifications

Quantity 1 g
UN Number 1759
Formula Weight 315.84
Percent Purity ≥97.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethynyl(phenyl)iodonium Tetrafluoroborate [Ethynylating Reagent]
TSCA No
Recommended Storage Freezer