missing translation for 'onlineSavingsMsg'
Learn More

Ethylurea 95.0+%, TCI America™

Catalog No. E019225G
Change view
Click to view available options
Quantity:
25 g
500 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
E019225G 25 g
E0192500G 500 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. E019225G Supplier TCI America Supplier No. E019225G

Chemical Identifiers

CAS 625-52-5
Molecular Formula C3H8N2O
Molecular Weight (g/mol) 88.11
MDL Number MFCD00007953
InChI Key RYECOJGRJDOGPP-UHFFFAOYSA-N
Synonym 1-ethylurea, n-ethylurea, urea, ethyl, urea, 1-ethyl, polyurethane, urea, n-ethyl, ethyl urea, pellethane, athylisoharnstoff
PubChem CID 12254
IUPAC Name ethylurea
SMILES CCNC(N)=O

Specifications

Melting Point 92°C
Color White-Yellow
Quantity 25 g
Formula Weight 88.11
Percent Purity ≥95.0% (T)
Physical Form Crystalline Powder
Chemical Name or Material Ethylurea
EINECSNumber (2)-2870
RTECSNumber YT2335000
TSCA Yes