accessibility menu, dialog, popup

UserName

CAS RN 625-52-5

IUPAC Name: ethylurea
Synonyms: ethylurea
Molecular Weight (g/mol): 88.11
Molecular Formula: C3H8N2O
InChi Key: RYECOJGRJDOGPP-UHFFFAOYSA-N
SMILES: CCNC(N)=O
Percent Purity 
  • (2)
  • (3)
Quantity 
  • (1)
  • (2)
  • (2)
Physical Form 
  • (2)

Filtered Search Results

Products from some of our suppliers do not display in filtered search results. Please clear all filters to see these products.

Narrow Results

Narrow Results

  • (3)
  • (2)
  • (3)
  • (2)
  • (3)
  • (1)
  • (2)
  • (2)
  • (2)